Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CGGK acetate | 96.78% | 363.43 (free acid) | 1 MG
SDP

Catalog No. 5000425588
Encompass_Preferred

CGGK acetate is a control peptide of CAQK peptide, intended for research use only. It presents as a white to off-white solid with a purity of 96.78% and a molecular weight of 363.43 (free acid). Its chemical formula is C13H25N5O5S.xC2H4O2. This product is shipped at room temperature and requires sealed storage, away from moisture.

  • Sequence: Cys-Gly-Gly-Lys
  • Purity: 96.78%
  • Solubility in DMSO: ≥ 100 mg/mL
  • Powder storage: -80°C for 2 years, or -20°C for 1 year
  • In solvent storage: -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-004-25588 Supplier Medchemexpress LLC Supplier No. HYP10737A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGGK acetate is a control peptide of CAQK peptide, intended for research use only. It presents as a white to off-white solid with a purity of 96.78% and a molecular weight of 363.43 (free acid). Its chemical formula is C13H25N5O5S.xC2H4O2. This product is shipped at room temperature and requires sealed storage, away from moisture.

  • Sequence: Cys-Gly-Gly-Lys
  • Purity: 96.78%
  • Solubility in DMSO: ≥ 100 mg/mL
  • Powder storage: -80°C for 2 years, or -20°C for 1 year
  • In solvent storage: -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.