Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CGGK acetate | 96.78% | 363.43 (free acid) | 5 MG
SDP

Catalog No. 5000425686
Encompass_Preferred

CGGK acetate is a control peptide of CAQK peptide. It is supplied as a white to off-white solid with a sequence of Cys-Gly-Gly-Lys. This peptide is soluble in DMSO at concentrations of 100 mg/mL or greater.

  • High purity of 96.78%
  • Molecular weight of 363.43 (free acid)
  • Appearance as a solid
  • Color is white to off-white
  • Soluble in DMSO (≥ 100 mg/mL)
  • Storage conditions: -80°C for 2 years or -20°C for 1 year (powder); -80°C for 6 months or -20°C for 1 month (in solvent)

Catalog No. 50-004-25686 Supplier Medchemexpress LLC Supplier No. HYP10737A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGGK acetate is a control peptide of CAQK peptide. It is supplied as a white to off-white solid with a sequence of Cys-Gly-Gly-Lys. This peptide is soluble in DMSO at concentrations of 100 mg/mL or greater.

  • High purity of 96.78%
  • Molecular weight of 363.43 (free acid)
  • Appearance as a solid
  • Color is white to off-white
  • Soluble in DMSO (≥ 100 mg/mL)
  • Storage conditions: -80°C for 2 years or -20°C for 1 year (powder); -80°C for 6 months or -20°C for 1 month (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.