Learn More
Medchemexpress LLC Cggk acetate | 96.8% | 363.43 | 10 MG
SDP
Catalog No. 5000425219
Encompass_Preferred
CGGK acetate is a control peptide for CAQK peptide. This peptide is provided as a solid, white to off-white appearance with a purity of 96.78%. It is suitable for various research applications.
  • For research use only
  • Ships at room temperature in continental US
  • Store sealed, away from moisture
  • Soluble in DMSO (≥ 100 mg/mL)
Catalog No. 50-004-25219 Supplier Medchemexpress LLC Supplier No. HYP10737A10MG
CGGK acetate is a control peptide for CAQK peptide. This peptide is provided as a solid, white to off-white appearance with a purity of 96.78%. It is suitable for various research applications.
  • For research use only
  • Ships at room temperature in continental US
  • Store sealed, away from moisture
  • Soluble in DMSO (≥ 100 mg/mL)