Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CGGRGD TFA | 98.7% | 677.61 | 5 MG
SDP

Catalog No. 5000423157
Encompass_Preferred

CGGRGD TFA is a derivative of RGD (Arginine-Glycine-Aspartic acid) that includes cysteine at its N-terminal. This product is intended for research applications only and not for human use. It is referenced in publications regarding the functionalization of PCL fibrous membranes with RGD peptide.

  • Biological activity: a derivative of RGD with cysteine at its N-terminal
  • Purity: 98.66%
  • Molecular weight: 677.61
  • Formula: C21H34F3N9O11S
  • Appearance: solid
  • Color: white to off-white
  • Sequence: Cys-Gly-Gly-Arg-Gly-Asp
  • Sequence shortening: CGGRGD
  • Solubility in vitro: soluble in DMSO at 100 mg/mL (147.58 mM); ultrasonic assistance may be needed, and newly opened DMSO is recommended due to hygroscopic impact on solubility

Catalog No. 50-004-23157 Supplier Medchemexpress LLC Supplier No. HYP2219A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGGRGD TFA is a derivative of RGD (Arginine-Glycine-Aspartic acid) that includes cysteine at its N-terminal. This product is intended for research applications only and not for human use. It is referenced in publications regarding the functionalization of PCL fibrous membranes with RGD peptide.

  • Biological activity: a derivative of RGD with cysteine at its N-terminal
  • Purity: 98.66%
  • Molecular weight: 677.61
  • Formula: C21H34F3N9O11S
  • Appearance: solid
  • Color: white to off-white
  • Sequence: Cys-Gly-Gly-Arg-Gly-Asp
  • Sequence shortening: CGGRGD
  • Solubility in vitro: soluble in DMSO at 100 mg/mL (147.58 mM); ultrasonic assistance may be needed, and newly opened DMSO is recommended due to hygroscopic impact on solubility

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.