Learn More
Enzo Life Sciences CGH labeling kit for BAC arrays (20Reactions)
SDP
Supplier:  Enzo Life Sciences ENZ42670
Encompass_Preferred
For preparation  of Cyanine 3- and Cyanine 5-labeled DNA for hybridization to BAC arrays.The CGH Labeling Kit for BAC Arrays has been optimized to label DNA for dual sample comparative genomic hybridization (CGH) assays on BAC arrays. All required reagents are provided including fully optimized nucleotide mixes that contain either cyanine 3-dUTP or cyanine 5-dUTP.The CGH Labeling Kit for BAC Arrays has been specifically tailored to meet the varying requirements of individual laboratories by providing sufficient amount of reagents for two different protocols. For the average user, the Standard Protocol calls for a 4-hour labeling in a 50µl reaction volume. For users with dilute DNA (≤ 20 ng/µl), the Dilute DNA Protocol calls for a 4-16 hour labeling in a total volume of 100µl. Applications: CGH.
Catalog No. 50-201-2530