Learn More
Aviva Systems Biology Corp CGI-143 Antibody - N-terminal region (ARP32073_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32073_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CGI-143. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CGI-143. Alternate names: hBolA, CGI-143, RP11-196G18.18ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54445