Learn More
Apexbio Technology LLC CGI-1746, 10mg. Cas: 910232-84-7 MFCD: MFCD18782602. Btk inhibitor
SDP
Supplier:  Apexbio Technology LLC A330210
Encompass_Preferred
CGI-1746 is a selective and potent inhibitor of Bruton's tyrosine kinase (BTK) with IC50 value of 1.9 nM [1][2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4794