Learn More
Aviva Systems Biology Corp CGI-62 Antibody - C-terminal region (ARP32751_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32751_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CGI-62. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CGI-62. Alternate names: CGI-62, C8orf70, FAM164AApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Rabbit, Rat
Catalog No. 50-014-52872