Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGK 733, Tocris Bioscience™

Catalog No. 263910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-391-0 10 mg
26-395-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-391-0 Supplier R&D Systems Supplier No. 2639/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Induces apoptosis in prematurely senescent cells

Originally defined as a selective inhibitor of ATR and ATM kinases. Induces cell death in prematurely senescent breast cancer cells. Decreases p21CIP1 levels in premature senescent MCF-7 and HCT-116 cells; also exhibits antiproliferative activity in a range of cancer cell lines. Blocks camptothecin -induced p53 phosphorylation and protects cells from camptothecin-induced apoptosis.

Chemical Identifiers

CAS 905973-89-9
Molecular Formula C23H18Cl3FN4O3S
Molecular Weight (g/mol) 555.83
InChI Key HLCDNLNLQNYZTK-UHFFFAOYSA-N
Synonym 2,2-diphenyl-n-2,2,2-trichloro-1-3-4-fluoro-3-nitrophenyl thioureido ethyl acetamide, 2,2-diphenyl-n-2,2,2-trichloro-1-4-fluoro-3-nitrophenyl carbamothioylamino ethyl acetamide, diphenyl acetamidotrichloroethyl fluoronitrophenyl thiourea, unii-l3dgz99qym, 2,2-diphenyl-n-2,2,2-trichloro-1-4-fluoro-3-nitrophenyl carbamothioyl amino ethyl acetamide, l3dgz99qym, d00pxo, diphenyl acetamidotrichloroethyl fluoronitrophenyl thiourea inci
PubChem CID 6605258
IUPAC Name 2,2-diphenyl-N-[2,2,2-trichloro-1-[(4-fluoro-3-nitrophenyl)carbamothioylamino]ethyl]acetamide
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)NC(C(Cl)(Cl)Cl)NC(=S)NC3=CC(=C(C=C3)F)[N+](=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 555.84
Percent Purity >98%
Chemical Name or Material CGK 733
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.