Learn More
Lonza Walkersville CGM Chondrocyte Growth Medium BulletKit
Supplier:  Lonza Walkersville CC3216
Encompass
Articular cartilage is characterized by high wear resistance and poor regenerative qualities. It is responsible for much of the compressive resistance and load bearing qualities of the knee joint.CGM Chondrocyte Growth Medium is specially designed for expansion of chondrocytes. The BulletKit is conveniently packaged with basal medium and supplements included for the growth of chondrocytes.CDM Chondrocyte Differentiation Medium is also available and sold separately for differentiation of human chondrocytes.Applications:Osteoarthritis Cartilage failureCellular matrix linkage ruptureChondrocyte protein synthesis inhibition Chondrocyte apoptosisContent:1 x Bottle Basal Medium 500 mL1 x Bottle FBS 25.00 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Yellow Cap Vial with R3-IGF 1.00 mL1 x Bottle bFGF 2.50 mL1 x Natural Cap Vial with Transferrin 0.50 mL1 x Lilac Cap Vial with Insulin 1.00 mL
Catalog No. NC9930470