Learn More
Eurofins Discoverx HitHunter cGMP Assay, 10,000 dp (384-well). (90-0039L)
Catalog No. 501999529
Encompass
The HitHunter cGMP Assay is a detection kit offering a robust, sensitive and easy-to-use assay to measure intracellular cGMP levels. This product uses EFC technology, and contains ED-tagged cGMP, anti-cGMP antibody, and pure EA. Upon application to a cell which contains endogenous, untagged cGMP, the anti-cGMP antibody will be sequestered by binding to this untagged cGMP, allowing for EFC between the tagged cGMP and soluble EA. This will produce a proportional level of chemiluminescent signal. This kit may be used with Eurofins DiscoverX cell lines as well as cell lines from other sources. This detection kit contains reagents for 25 plates (384 well). For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.
Catalog No. 50-199-9529 Supplier Eurofins Discoverx Supplier No. 900039L
Add to Cart
Add to Cart
Request a Quote
The HitHunter cGMP Assay is a detection kit offering a robust, sensitive and easy-to-use assay to measure intracellular cGMP levels. This product uses EFC technology, and contains ED-tagged cGMP, anti-cGMP antibody, and pure EA. Upon application to a cell which contains endogenous, untagged cGMP, the anti-cGMP antibody will be sequestered by binding to this untagged cGMP, allowing for EFC between the tagged cGMP and soluble EA. This will produce a proportional level of chemiluminescent signal. This kit may be used with Eurofins DiscoverX cell lines as well as cell lines from other sources. This detection kit contains reagents for 25 plates (384 well). For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.