Learn More
Eurofins Discoverx HitHunter cGMP Assay, 2,400 dp (384-well). (90-0039M)
Catalog No. 501999530
Encompass
The HitHunter cGMP Assay is a detection kit offering a robust, sensitive and easy-to-use assay to measure intracellular cGMP levels. This product uses EFC technology, and contains ED-tagged cGMP, anti-cGMP antibody, and pure EA. Upon application to a cell which contains endogenous, untagged cGMP, the anti-cGMP antibody will be sequestered by binding to this untagged cGMP, allowing for EFC between the tagged cGMP and soluble EA. This will produce a proportional level of chemiluminescent signal. This kit may be used with Eurofins DiscoverX cell lines as well as cell lines from other sources. This detection kit contains reagents for 6 plates (384 well). For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.
Catalog No. 50-199-9530 Supplier Eurofins Discoverx Supplier No. 900039M
The HitHunter cGMP Assay is a detection kit offering a robust, sensitive and easy-to-use assay to measure intracellular cGMP levels. This product uses EFC technology, and contains ED-tagged cGMP, anti-cGMP antibody, and pure EA. Upon application to a cell which contains endogenous, untagged cGMP, the anti-cGMP antibody will be sequestered by binding to this untagged cGMP, allowing for EFC between the tagged cGMP and soluble EA. This will produce a proportional level of chemiluminescent signal. This kit may be used with Eurofins DiscoverX cell lines as well as cell lines from other sources. This detection kit contains reagents for 6 plates (384 well). For detailed product information, visit www.discoverx.com/products-applications/biochemical-assays.