Learn More
Alpco Ltd cGMP Complete ELISA 5 Plate
SDP
Supplier:  Alpco Ltd 74CGMHUE05
Encompass
The cGMP Complete ELISA 5 Plate Kit is for the quantitative determination of cGMP in cell lysates, culture supernatants, plasma, serum, saliva, tissue, and urine from any species.
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2332825
 Restricted Item