Learn More
STEMCELL Technologies Gentle Cell Dissociation Reagent, Size: 100 mL
SDP
Catalog No. 502069325
Encompass
Gentle Cell Dissociation Reagent (GCDR) is an enzyme-free reagent suitable for the dissociation of human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells into cell aggregates for routine passaging or into a single-cell suspension. It is also suitable for the isolation of intestinal crypts to establish intestinal organoids, and for breaking up Corning Matrigel domes when passaging organoid cultures. GCDR does not contain enzymes or other proteins.GCDR is now also manufactured following relevant cGMPs under a certified quality management system to ensure the highest quality and consistency for reproducible results.To request a Letter of Authorization (LOA) for Gentle Cell Dissociation Reagent’s Drug Master File, click here.
Catalog No. 50-206-9325 Supplier STEMCELL Technologies Supplier No. 1000485
Gentle Cell Dissociation Reagent (GCDR) is an enzyme-free reagent suitable for the dissociation of human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells into cell aggregates for routine passaging or into a single-cell suspension. It is also suitable for the isolation of intestinal crypts to establish intestinal organoids, and for breaking up Corning Matrigel domes when passaging organoid cultures. GCDR does not contain enzymes or other proteins.GCDR is now also manufactured following relevant cGMPs under a certified quality management system to ensure the highest quality and consistency for reproducible results.To request a Letter of Authorization (LOA) for Gentle Cell Dissociation Reagent’s Drug Master File, click here.