Learn More
Akron Biotechnology LLC Heparin Sodium Salt, GMP Grade, 1 g Bottle, AK9987-1000
Catalog No. NC2159098
Encompass
Akron’s Heparin Sodium Salt is manufactured following relevant cGMP guidelinesProduct Features:Raw Material•Country of Origin: Brazil•Inspected and registered animal facilities•Sanitary Certificate supporting each batch of raw materialManufacturing•Type II eCTD MF (#026153) on file with FDA•Validated alkaline and heat treatment for inactivation of conventional viruses•Validated removal of protein impurities and pigments•Sterile microfiltration before dry milling and homogenizationStability•5-year shelf life•Store at or below 40 °C•Protect from light and humidity•Transport ambient temperatureFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
This item is not returnable. View return policy
Catalog No. NC2159098 Supplier Akron Biotechnology LLC Supplier No. AK99871000
Akron’s Heparin Sodium Salt is manufactured following relevant cGMP guidelinesProduct Features:Raw Material•Country of Origin: Brazil•Inspected and registered animal facilities•Sanitary Certificate supporting each batch of raw materialManufacturing•Type II eCTD MF (#026153) on file with FDA•Validated alkaline and heat treatment for inactivation of conventional viruses•Validated removal of protein impurities and pigments•Sterile microfiltration before dry milling and homogenizationStability•5-year shelf life•Store at or below 40 °C•Protect from light and humidity•Transport ambient temperatureFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.