Learn More
Enzo Life Sciences cGMP polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADI915063100
Encompass_Preferred
Host: Rabbit. Source: Whole rabbit serum. Species reactivity: Species independent. Applications: ELISA. Recommended Dilutions: EIA (1:4,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Contains 0.1% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7108