Learn More
STEMCELL Technologies mTeSR™ Plus
SDP
Catalog No. 502036627
Encompass
Enjoy weekend-free schedules and enhanced growth characteristics while maintaining cell quality with stabilized feeder-free maintenance medium for human pluripotent stem cells (hPSCs). Manufactured under relevant cGMPs, mTeSR™ Plus supports fundamental research as well as cell therapy and IND research applications. Based on mTeSR™1 (Catalog #85850), with stabilized critical medium components, including FGF2, and enhanced pH buffering, cell-quality attributes can be maintained with daily or restricted feeding. Each lot of mTeSR™ Plus 5X Supplement is used to prepare complete mTeSR™ Plus medium and performance-tested in hPSC culture assays.This product is compatible with a variety of matrices, including Corning Matrigel hESC-Qualified Matrix and Vitronectin XF™ (Catalog #07180, developed and manufactured by Nucleus Biologics).
This item is not returnable. View return policy
Catalog No. 50-203-6627 Supplier STEMCELL Technologies Supplier No. 1000276
Enjoy weekend-free schedules and enhanced growth characteristics while maintaining cell quality with stabilized feeder-free maintenance medium for human pluripotent stem cells (hPSCs). Manufactured under relevant cGMPs, mTeSR™ Plus supports fundamental research as well as cell therapy and IND research applications. Based on mTeSR™1 (Catalog #85850), with stabilized critical medium components, including FGF2, and enhanced pH buffering, cell-quality attributes can be maintained with daily or restricted feeding. Each lot of mTeSR™ Plus 5X Supplement is used to prepare complete mTeSR™ Plus medium and performance-tested in hPSC culture assays.This product is compatible with a variety of matrices, including Corning Matrigel hESC-Qualified Matrix and Vitronectin XF™ (Catalog #07180, developed and manufactured by Nucleus Biologics).