Learn More
Lonza Walkersville CGM Chondrocyte Growth Medium SingleQuots Supplements and Growth Factors
Catalog No. NC1925829
Encompass
CGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the CGM Chondrocyte Growth Medium BulletKit (Catalog Number CC-3216). The full CGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3216.CGM Chondrocyte Growth Medium is specially designed for expansion of chondrocytes. The BulletKit is conveniently packaged with basal medium and supplements included for the growth of chondrocytes.CDM Chondrocyte Differentiation Medium is also available and sold separately for differentiation of human chondrocytes.Content:1 x Bottle FBS 25.00 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Yellow Cap Vial with R3-IGF 1.00 mL1 x Bottle bFGF 2.50 mL1 x Natural Cap Vial with Transferrin 0.50 mL1 x Lilac Cap Vial with Insulin 1.00 mL
Catalog No. NC1925829 Supplier Lonza Walkersville Supplier No. CC4409
CGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the CGM Chondrocyte Growth Medium BulletKit (Catalog Number CC-3216). The full CGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3216.CGM Chondrocyte Growth Medium is specially designed for expansion of chondrocytes. The BulletKit is conveniently packaged with basal medium and supplements included for the growth of chondrocytes.CDM Chondrocyte Differentiation Medium is also available and sold separately for differentiation of human chondrocytes.Content:1 x Bottle FBS 25.00 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Yellow Cap Vial with R3-IGF 1.00 mL1 x Bottle bFGF 2.50 mL1 x Natural Cap Vial with Transferrin 0.50 mL1 x Lilac Cap Vial with Insulin 1.00 mL