Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cgp 12177 hydrochloride | 64208-32-8 | 99.7% | 315.80 g·mol⁻¹ | C14H22ClN3O3 | 5 MG
SDP

Catalog No. 5000211767
Encompass_Preferred

CGP 12177 hydrochloride is a research-grade β-adrenergic receptor ligand supplied as the hydrochloride salt. It is used in pharmacological studies of β-adrenoceptors and in cardiovascular and metabolic disease research.

  • High reported purity (~99.7%).
  • Molecular formula C14H22ClN3O3 and molecular weight 315.80 g·mol⁻¹.
  • Acts as a β3-adrenergic receptor agonist and β1/β2 antagonist in receptor assays.
  • Available in small milligram pack sizes suitable for laboratory studies.
  • Provided with supporting datasheet and specifications for research use.

Catalog No. 50-002-11767 Supplier Medchemexpress LLC Supplier No. HY101393A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGP 12177 hydrochloride is a research-grade β-adrenergic receptor ligand supplied as the hydrochloride salt. It is used in pharmacological studies of β-adrenoceptors and in cardiovascular and metabolic disease research.

  • High reported purity (~99.7%).
  • Molecular formula C14H22ClN3O3 and molecular weight 315.80 g·mol⁻¹.
  • Acts as a β3-adrenergic receptor agonist and β1/β2 antagonist in receptor assays.
  • Available in small milligram pack sizes suitable for laboratory studies.
  • Provided with supporting datasheet and specifications for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.