Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cgp 20712 mesylate | 105737-62-0 | MFCD00798870 | 99.0% | 590.57 | C24H29F3N4O8S | 5 MG
SDP

Catalog No. 5000212614
Encompass_Preferred

CGP 20712 A (mesylate) is a selective β1-adrenoceptor antagonist intended for research use in receptor pharmacology and biochemical assays. It is provided as a high-purity solid for in-vitro binding and functional studies, offering reliable potency and selectivity for β1 receptor profiling.

  • Selective β1-adrenoceptor antagonist with reported IC50 of 0.7 nM.
  • Approximately 10,000-fold selectivity over β2-adrenoceptors.
  • High purity (99.0%) suitable for analytical and research applications.
  • Molecular weight 590.57 and molecular formula C24H29F3N4O8S.
  • Supplied as a solid; recommended storage at -20°C, sealed and away from moisture.
  • Small package sizes appropriate for assay development and reference use (5 MG).

Catalog No. 50-002-12614 Supplier Medchemexpress LLC Supplier No. HY101355B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGP 20712 A (mesylate) is a selective β1-adrenoceptor antagonist intended for research use in receptor pharmacology and biochemical assays. It is provided as a high-purity solid for in-vitro binding and functional studies, offering reliable potency and selectivity for β1 receptor profiling.

  • Selective β1-adrenoceptor antagonist with reported IC50 of 0.7 nM.
  • Approximately 10,000-fold selectivity over β2-adrenoceptors.
  • High purity (99.0%) suitable for analytical and research applications.
  • Molecular weight 590.57 and molecular formula C24H29F3N4O8S.
  • Supplied as a solid; recommended storage at -20°C, sealed and away from moisture.
  • Small package sizes appropriate for assay development and reference use (5 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.