Learn More
TARGETMOL CHEMICALS INC CGP 3466B MALEATE 5MG
SDP
Catalog No. 502581002
Encompass_Preferred
Also available in 2 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CGP 3466B maleate (Omigapil maleate) is an orally bioavailable GAPDH nitrosylation inhibitor. Omigapil maleate abrogates A(beta)1-42-induced tau acetylation memory impairment and locomotor dysfunction in mice. Omigapil maleate has the potential for the research of Alzheimer's disease. Omigapil maleate (CGP3446B maleate) is a apoptosis inhibitor. Omigapil maleate can be used for the research of congenital muscular dystrophy (CMD). purity: 98%
Catalog No. 50-258-1002 Supplier TARGETMOL CHEMICALS INC Supplier No. T217925MG
Also available in 2 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CGP 3466B maleate (Omigapil maleate) is an orally bioavailable GAPDH nitrosylation inhibitor. Omigapil maleate abrogates A(beta)1-42-induced tau acetylation memory impairment and locomotor dysfunction in mice. Omigapil maleate has the potential for the research of Alzheimer's disease. Omigapil maleate (CGP3446B maleate) is a apoptosis inhibitor. Omigapil maleate can be used for the research of congenital muscular dystrophy (CMD). purity: 98%