Learn More
Cayman Chemical CGP 36216hydrochlorIde 10mg
SDP
Catalog No. 502953933
Encompass_Preferred
A selective antagonist of GABAB receptors (IC50 = 43 µM); active at presynaptic, but not postsynaptic, receptors
Catalog No. 50-295-3933 Supplier Cayman Chemical Supplier No. 1198510 MG
A selective antagonist of GABAB receptors (IC50 = 43 µM); active at presynaptic, but not postsynaptic, receptors