Learn More
Abcam CGP 37157, Na+-Ca2+ e x change inhibitor, 10MG
Catalog No. 502707942
Encompass_Preferred
MW 324.2 Da, Purity >99%. Selective inhibitor of the mitochondrial Na+-Ca2+ exchanger (IC50 = 0.4 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7942 Supplier Abcam Supplier No. AB12001210MG
MW 324.2 Da, Purity >99%. Selective inhibitor of the mitochondrial Na+-Ca2+ exchanger (IC50 = 0.4 μM).
The product is subject to the following: Abcam Restricted Use Statement