Learn More
Medchemexpress LLC HY-107702 5mg Medchemexpress, CGP 37849 CAS:127910-31-0 Purity:>98%
SDP
Catalog No. 501968455
Encompass_Preferred
Medchemexpress, HY-107702 5mg CGP 37849 CAS:127910-31-0 CGP 37849 is a potent, competitive and orally active N-methyl-D-aspartate (NMDA) receptor antagonist. CGP 37849 is an anticonvulsant in rodents and has antidepressant and anxiolytic-like effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8455 Supplier Medchemexpress LLC Supplier No. HY1077025MG
Medchemexpress, HY-107702 5mg CGP 37849 CAS:127910-31-0 CGP 37849 is a potent, competitive and orally active N-methyl-D-aspartate (NMDA) receptor antagonist. CGP 37849 is an anticonvulsant in rodents and has antidepressant and anxiolytic-like effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.