Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CGP 44 645 | 134521-16-7 | 234.25 | 100 MG
SDP

Catalog No. 5000447455
Encompass_Preferred

CGP 44 645 is the methanol metabolite of Letrozole and serves as a molecular indicator for evaluating its metabolic kinetics in microsomes in vitro. Levels in rat liver microsomes may be sex-specific. This product is for research use only.

  • Purity: 98.1%
  • Molecular weight: 234.25
  • Formula: C15H10N2O
  • Appearance: Solid, white to off-white
  • Solubility in DMSO: ≥ 200 mg/mL (853.79 mM)
  • Recommended storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-47455 Supplier Medchemexpress LLC Supplier No. HYW003371100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGP 44 645 is the methanol metabolite of Letrozole and serves as a molecular indicator for evaluating its metabolic kinetics in microsomes in vitro. Levels in rat liver microsomes may be sex-specific. This product is for research use only.

  • Purity: 98.1%
  • Molecular weight: 234.25
  • Formula: C15H10N2O
  • Appearance: Solid, white to off-white
  • Solubility in DMSO: ≥ 200 mg/mL (853.79 mM)
  • Recommended storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.