Learn More
Medchemexpress LLC CGP 54626 hydrochloride | 149184-21-4 | 99.8% | 444.76 g/mol | C18H29Cl3NO3P | 1 MG
SDP
Catalog No. 5000213221
Encompass_Preferred
CGP 54626 hydrochloride is a selective GABAB receptor antagonist for research use. It is supplied as a white to off-white solid with high purity and defined molecular properties, and is commonly used in receptor-binding and functional assays to study GABAB-mediated signaling.
  • Selective GABAB receptor antagonist (IC50 ≈ 4 nM).
  • High purity (≈99.8%).
  • White to off-white solid appearance.
  • Recommended storage: protected from light at -20°C; in solution -80°C for long term.
  • Suitable for in vitro binding and functional assays.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-13221 Supplier Medchemexpress LLC Supplier No. HY1013781MG
CGP 54626 hydrochloride is a selective GABAB receptor antagonist for research use. It is supplied as a white to off-white solid with high purity and defined molecular properties, and is commonly used in receptor-binding and functional assays to study GABAB-mediated signaling.
  • Selective GABAB receptor antagonist (IC50 ≈ 4 nM).
  • High purity (≈99.8%).
  • White to off-white solid appearance.
  • Recommended storage: protected from light at -20°C; in solution -80°C for long term.
  • Suitable for in vitro binding and functional assays.
  • For research use only; not for human or veterinary use.