Learn More
Cayman Chemical CGP 54626hydrochlorIde 10mg
SDP
Catalog No. 502983657
Encompass_Preferred
A selective GABAB receptor antagonist (IC50 = 4 nM)
Catalog No. 50-298-3657 Supplier Cayman Chemical Supplier No. 2012110 MG
A selective GABAB receptor antagonist (IC50 = 4 nM)