Learn More
Cayman Chemical CGP 54626hydrochlorIde 50mg
SDP
Catalog No. 502982768
Encompass_Preferred
A selective GABAB receptor antagonist (IC50 = 4 nM)
Catalog No. 50-298-2768 Supplier Cayman Chemical Supplier No. 2012150 MG
A selective GABAB receptor antagonist (IC50 = 4 nM)