Learn More
Abcam CGP 55845 hydrochloride, GABAB antagonist, 50MG
Catalog No. 502584621
Encompass_Preferred
MW 438.7 Da, Purity >99%. Potent, selective GABAB antagonist (IC50 = 5 nM). Prevents agonist binding (pKi = 8.35) and blocks GABAB responses to Baclofen (ab120325). Inhibits GABA and glutamate release (pEC50 values are 8.08 and 7.85, respectively). Stimulates glucose induced insulin secretion.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4621 Supplier Abcam Supplier No. AB12033750MG
MW 438.7 Da, Purity >99%. Potent, selective GABAB antagonist (IC50 = 5 nM). Prevents agonist binding (pKi = 8.35) and blocks GABAB responses to Baclofen (ab120325). Inhibits GABA and glutamate release (pEC50 values are 8.08 and 7.85, respectively). Stimulates glucose induced insulin secretion.
The product is subject to the following: Abcam Restricted Use Statement