Learn More
Apexbio Technology LLC CGP 57380 522629-08-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063909
Encompass_Preferred
CGP 57380 (CAS 522629-08-9) is a specific and selective inhibitor targeting the serine/threonine kinase MNK1 an enzyme implicated in translation regulation via phosphorylation of the eukaryotic initiation factor 4E (eIF4E) It inhibits MNK1 activity in vitro with an IC50 of approximately 2 2 M CGP 57380 suppresses stimulus-induced phosphorylation of eIF4E in various cellular contexts including 293 cells vascular smooth muscle cells and macrophages reducing translation of mRNAs encoding proinflammatory mediators such as TNF- IL-6 and monocyte chemoattractant protein-1 (MCP-1) Hence CGP 57380 is valuable for investigating MNK1-mediated translational control in inflammation and cell signaling
Catalog No. 50-000-63909 Supplier Apexbio Technology LLC Supplier No. B35705.1
 Restricted Item
Add to Cart
Add to Cart
CGP 57380 (CAS 522629-08-9) is a specific and selective inhibitor targeting the serine/threonine kinase MNK1 an enzyme implicated in translation regulation via phosphorylation of the eukaryotic initiation factor 4E (eIF4E) It inhibits MNK1 activity in vitro with an IC50 of approximately 2 2 M CGP 57380 suppresses stimulus-induced phosphorylation of eIF4E in various cellular contexts including 293 cells vascular smooth muscle cells and macrophages reducing translation of mRNAs encoding proinflammatory mediators such as TNF- IL-6 and monocyte chemoattractant protein-1 (MCP-1) Hence CGP 57380 is valuable for investigating MNK1-mediated translational control in inflammation and cell signaling