Learn More
Apexbio Technology LLC CGP 57380, 5mg. Cas: 522629-08-9 MFCD: MFCD03861062. MNK1 inhibitor, specific and selective
SDP
Supplier:  Apexbio Technology LLC B35705
Encompass_Preferred
CGP 57380 is a specific and selective inhibitor of MNK1 with IC50 value of 2.2 M [1]. Mitogen-activated protein (MAP) kinase interacting kinases 1 (MNK1) is a serine/threonine kinase and is able to integrate signal from MAP kinase pathway and phosphorylate eIF4E [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5347