Learn More
Cayman Chemical CGP 77675hydrate 10mg
SDP
Catalog No. 502983109
Encompass_Preferred
An inhibitor of Src family kinases that blocks the phosphorylation of peptide substrates and autophosphorylation of purified c-Src (IC50s = 5-20 and 40 nM, respectively); also inhibits Lck and c-Yes with low nanomolar IC50 values
Catalog No. 50-298-3109 Supplier Cayman Chemical Supplier No. 2108910 MG
An inhibitor of Src family kinases that blocks the phosphorylation of peptide substrates and autophosphorylation of purified c-Src (IC50s = 5-20 and 40 nM, respectively); also inhibits Lck and c-Yes with low nanomolar IC50 values