Learn More
Medchemexpress LLC CGP 78608 hydrochlor 1mg | 1135278-54-4 | 1MG
SDP
Catalog No. 5000241695
Encompass_Preferred
CGP 78608 hydrochloride is a highly potent and selective antagonist at the glycine-binding site of the NMDA receptor with an IC50 of 6 nM CGP 78608 hydrochloride acts as a potentiator of GluN1/GluN3A-mediated glycine currents with an estimated EC50 in the low nM range (26 3 nM) Anticonvulsant activity[1 [2
Catalog No. 50-002-41695 Supplier Medchemexpress LLC Supplier No. HY1077011MG
CGP 78608 hydrochloride is a highly potent and selective antagonist at the glycine-binding site of the NMDA receptor with an IC50 of 6 nM CGP 78608 hydrochloride acts as a potentiator of GluN1/GluN3A-mediated glycine currents with an estimated EC50 in the low nM range (26 3 nM) Anticonvulsant activity[1 [2