Learn More
American Radiolabeled Chemicals Inc CGP 7930 3H 50UC
SDP
Catalog No. 50352914
Encompass_Preferred
CGP 7930-3H 50uCi in ethanol, 1 mCi/ml, 20-30 Ci/mmol, M.W. 292.46, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-914 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 193450UC
CGP 7930-3H 50uCi in ethanol, 1 mCi/ml, 20-30 Ci/mmol, M.W. 292.46, Shipped in Dry Ice, Exclusive