Learn More
Abcam CGP 7930, Positive allosteric modulator of GABAB receptors, 10MG
Supplier:  Abcam AB12049210MG
Encompass_Preferred
MW 292.5 Da, Purity >98%. Positive GABAB receptor allosteric modulator (EC50 values are 5.37 and 4.60 μM, respectively). Increases receptor agonist affinity and signal transduction efficacy. Also shown to potentiate receptor response to baclofen (ab120325).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7555