Learn More
Abcam CGP 7930, Positive allosteric modulator of GABAB receptors, 50MG
Catalog No. 502707554
Encompass_Preferred
MW 292.5 Da, Purity >98%. Positive GABAB receptor allosteric modulator (EC50 values are 5.37 and 4.60 μM, respectively). Increases receptor agonist affinity and signal transduction efficacy. Also shown to potentiate receptor response to baclofen (ab120325).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7554 Supplier Abcam Supplier No. AB12049250MG
MW 292.5 Da, Purity >98%. Positive GABAB receptor allosteric modulator (EC50 values are 5.37 and 4.60 μM, respectively). Increases receptor agonist affinity and signal transduction efficacy. Also shown to potentiate receptor response to baclofen (ab120325).
The product is subject to the following: Abcam Restricted Use Statement