Learn More
Medchemexpress LLC HY-15754 5mg Medchemexpress, CGP37157 CAS:75450-34-9 Purity:>98%
SDP
Catalog No. 502081793
Encompass_Preferred
Medchemexpress, HY-15754 5mg CGP37157 CAS:75450-34-9 CGP37157 is a potent, selective inhibitor of Na+/Ca2+ exchanger, inhibiting the Na+-induced Ca2+-release from guinea-pig heart mitochondria, with an IC50 of 0.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1793 Supplier Medchemexpress LLC Supplier No. HY157545MG
Medchemexpress, HY-15754 5mg CGP37157 CAS:75450-34-9 CGP37157 is a potent, selective inhibitor of Na+/Ca2+ exchanger, inhibiting the Na+-induced Ca2+-release from guinea-pig heart mitochondria, with an IC50 of 0.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.