Learn More
TARGETMOL CHEMICALS INC CGP52411 5MG
SDP
Catalog No. 502581666
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CGP52411 is an orally active and ATP-competitive inhibitor of EGFR (IC50 0.3 uM). CGP52411 blocks the toxic influx of Ca2+ ions into neuronal cells and dramatically inhibits and reverses the formation of (beta)-amyloid A(beta)42 fibril aggregates. CGP52411 can be used in studies about Alzheimer's diseases. purity: 99%
Catalog No. 50-258-1666 Supplier TARGETMOL CHEMICALS INC Supplier No. T107825MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CGP52411 is an orally active and ATP-competitive inhibitor of EGFR (IC50 0.3 uM). CGP52411 blocks the toxic influx of Ca2+ ions into neuronal cells and dramatically inhibits and reverses the formation of (beta)-amyloid A(beta)42 fibril aggregates. CGP52411 can be used in studies about Alzheimer's diseases. purity: 99%