Learn More
Medchemexpress LLC HY-103531 5mg Medchemexpress, CGP52432 CAS:139667-74-6 Purity:>98%
SDP
Catalog No. 501968125
Encompass_Preferred
Medchemexpress, HY-103531 5mg CGP52432 CAS:139667-74-6 CGP52432 is a GABAB receptor antagonist, with an IC50 of 85 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8125 Supplier Medchemexpress LLC Supplier No. HY1035315MG
Medchemexpress, HY-103531 5mg CGP52432 CAS:139667-74-6 CGP52432 is a GABAB receptor antagonist, with an IC50 of 85 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.