Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cgp55845 hydrochloride | 149184-22-5 | MFCD09266221 | 99.1% | 438.71 g/mol | C18H23Cl3NO3P | 5 MG
SDP

Catalog No. 5000248645
Encompass_Preferred

CGP55845 hydrochloride is the hydrochloride salt of CGP55845, a potent and selective GABAB receptor antagonist with low-nanomolar potency (IC50 ≈ 5-6 nM). It is supplied as a solid for research use in receptor pharmacology and neurological studies.

  • Potent GABAB receptor antagonist with low-nanomolar activity.
  • High purity (99.1%) suitable for research applications.
  • Molecular weight 438.71 g/mol, well characterized.
  • CAS number 149184-22-5 for unambiguous identification.
  • Provided as the hydrochloride salt in a 5 mg pack for small-scale experiments.

Catalog No. 50-002-48645 Supplier Medchemexpress LLC Supplier No. HY1035165MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGP55845 hydrochloride is the hydrochloride salt of CGP55845, a potent and selective GABAB receptor antagonist with low-nanomolar potency (IC50 ≈ 5-6 nM). It is supplied as a solid for research use in receptor pharmacology and neurological studies.

  • Potent GABAB receptor antagonist with low-nanomolar activity.
  • High purity (99.1%) suitable for research applications.
  • Molecular weight 438.71 g/mol, well characterized.
  • CAS number 149184-22-5 for unambiguous identification.
  • Provided as the hydrochloride salt in a 5 mg pack for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.