Learn More
Medchemexpress LLC HY-11009 10mg , CGP60474 CAS:164658-13-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1100910MG
Encompass_Preferred
HY-11009 10mg CGP60474 CAS: 164658-13-3 CGP60474, a highly potent anti-endotoxemic agent, is a potent cyclin-dependent kinase (CDK) inhibitor (IC50 values are 26, 3, 4, 216, 10, 200 and 13 nM for CDK1/B, CDK2/E, CDK2/A, CDK4/D, CDK5/p25, CDK7/H and CDK9/T, respectively). CGP60474 is a selective and ATP-competitive PKC inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9453