Learn More
Medchemexpress LLC HY-107723 5mg Medchemexpress, CGP71683 hydrochloride CAS:192322-50-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1077235MG
Encompass_Preferred
Medchemexpress, HY-107723 5mg CGP71683 HCl CAS:192322-50-2 CGP71683 hydrochloride is a competitive neuropeptide Y5 receptor antagonist with a Ki of 1.3 nM, and shows no obvious activity at Y1 receptor (Ki, >4000 nM) and Y2 receptor (Ki, 200 nM) in cell membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8457