Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CGP71683 hydrochloride | 192322-50-2 | MFCD07783999 | 99.4% | 512.07 | C26H30ClN5O2S | 10 MG
SDP

Catalog No. 5000240329
Encompass_Preferred

CGP71683 hydrochloride is the hydrochloride salt of a selective neuropeptide Y Y5 receptor antagonist used as a research tool in neuroscience and endocrinology. It is supplied as a high-purity solid for non-clinical laboratory use, with supporting datasheet and certificate of analysis available.

  • Selective antagonist for the neuropeptide Y Y5 receptor (Ki ~1.3 nM).
  • High reported purity, 99.4%.
  • Molecular weight 512.07 g/mol; chemical formula C26H30ClN5O2S.
  • CAS number 192322-50-2 for unambiguous identification.
  • Solid powder suitable for dissolution in appropriate solvents for assays.
  • Supplied with datasheet and certificate of analysis for quality assurance.
  • Available in small milligram pack sizes for screening and mechanistic studies.

Catalog No. 50-002-40329 Supplier Medchemexpress LLC Supplier No. HY10772310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGP71683 hydrochloride is the hydrochloride salt of a selective neuropeptide Y Y5 receptor antagonist used as a research tool in neuroscience and endocrinology. It is supplied as a high-purity solid for non-clinical laboratory use, with supporting datasheet and certificate of analysis available.

  • Selective antagonist for the neuropeptide Y Y5 receptor (Ki ~1.3 nM).
  • High reported purity, 99.4%.
  • Molecular weight 512.07 g/mol; chemical formula C26H30ClN5O2S.
  • CAS number 192322-50-2 for unambiguous identification.
  • Solid powder suitable for dissolution in appropriate solvents for assays.
  • Supplied with datasheet and certificate of analysis for quality assurance.
  • Available in small milligram pack sizes for screening and mechanistic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.