Learn More
Medchemexpress LLC CGP71683 hydrochloride | 192322-50-2 | MFCD07783999 | 99.4% | 512.07 g/mol | C26H30ClN5O2S | 1 ML
SDP
Catalog No. 5000240328
Encompass_Preferred
CGP71683 hydrochloride is a selective, non-peptide neuropeptide Y (NPY) Y5 receptor antagonist (Ki = 1.3 nM) commonly used in receptor pharmacology and signaling research. It demonstrates substantially reduced activity at Y1 and Y2 receptors and is supplied both as a solid and as a pre-made DMSO solution for ready-to-use assays.
  • Selective NPY Y5 receptor antagonist (Ki = 1.3 nM).
  • High selectivity over Y1 and Y2 receptors.
  • Available as solid or 10 mM solution in DMSO, 1 mL.
  • High purity suitable for research applications.
  • Intended for receptor pharmacology and signaling studies.
Catalog No. 50-002-40328 Supplier Medchemexpress LLC Supplier No. HY10772310MM 1ML
CGP71683 hydrochloride is a selective, non-peptide neuropeptide Y (NPY) Y5 receptor antagonist (Ki = 1.3 nM) commonly used in receptor pharmacology and signaling research. It demonstrates substantially reduced activity at Y1 and Y2 receptors and is supplied both as a solid and as a pre-made DMSO solution for ready-to-use assays.
  • Selective NPY Y5 receptor antagonist (Ki = 1.3 nM).
  • High selectivity over Y1 and Y2 receptors.
  • Available as solid or 10 mM solution in DMSO, 1 mL.
  • High purity suitable for research applications.
  • Intended for receptor pharmacology and signaling studies.