Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CGP71683 hydrochloride | 192322-50-2 | MFCD07783999 | 99.4% | 512.07 g/mol | C26H30ClN5O2S | 50 MG
SDP

Catalog No. 5000242402
Encompass_Preferred

CGP71683 hydrochloride is a selective, non-peptide neuropeptide Y (NPY) Y5 receptor antagonist used for in vitro pharmacology and receptor signaling studies. The compound is supplied as a white solid and as a 10 mM solution in DMSO and is reported with high purity for research applications.

  • Selective NPY Y5 receptor antagonist with low nanomolar affinity.
  • Reported high purity suitable for research use.
  • Available as a solid and as a 10 mM solution in DMSO for assay convenience.
  • Molecular weight 512.07 g/mol aiding stoichiometric calculations.
  • CAS registry number 192322-50-2 for unambiguous identification.
  • Intended for biochemical and pharmacological studies in vitro.

Catalog No. 50-002-42402 Supplier Medchemexpress LLC Supplier No. HY10772350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CGP71683 hydrochloride is a selective, non-peptide neuropeptide Y (NPY) Y5 receptor antagonist used for in vitro pharmacology and receptor signaling studies. The compound is supplied as a white solid and as a 10 mM solution in DMSO and is reported with high purity for research applications.

  • Selective NPY Y5 receptor antagonist with low nanomolar affinity.
  • Reported high purity suitable for research use.
  • Available as a solid and as a 10 mM solution in DMSO for assay convenience.
  • Molecular weight 512.07 g/mol aiding stoichiometric calculations.
  • CAS registry number 192322-50-2 for unambiguous identification.
  • Intended for biochemical and pharmacological studies in vitro.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.