Learn More
Medchemexpress LLC HY-W062835 5mg Medchemexpress, CGP77675 CAS:234772-64-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0628355MG
Encompass_Preferred
Medchemexpress, HY-W062835 5mg CGP77675 CAS:234772-64-6 CGP77675 is a potent inhibitor of Src family kinases. CGP77675 inhibits Src, EGFR, KDR, v-Abl, and Lck with IC50s of 0.02, 0.15, 1.0, 0.31, and 0.29 μM, respectively. Anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3495