Learn More
Cayman Chemical CGRP AffInIty Sorbent 1mL
SDP
Catalog No. 502894812
Encompass_Preferred
The CGRP affinity sorbent contains immobilized mouse anti-CGRP. This product is designed for use in preparing CGRP-free plasma, which can be used as a sample matrix blank (SMB) for diluting standards and samples prior to analysis by EIA.
Catalog No. 50-289-4812 Supplier Cayman Chemical Supplier No. 4890091 ML
The CGRP affinity sorbent contains immobilized mouse anti-CGRP. This product is designed for use in preparing CGRP-free plasma, which can be used as a sample matrix blank (SMB) for diluting standards and samples prior to analysis by EIA.