Learn More
Medchemexpress LLC HY-P1548A 1mg Medchemexpress, β-CGRP, human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1548A1MG
Encompass_Preferred
Medchemexpress, HY-P1548A 1mg β-CGRP, human (TFA) CAS: β-CGRP, human TFA is one of calcitonin peptides, acts via the complex of calcitonin-receptor-like receptor (CRLR) and receptor-activity-modifying protein (RAMP), with IC 50 s of 1 nM and 300 nM for CRLR/RAMP1 and CRLR/RAMP2 in cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4224