Learn More
Medchemexpress LLC α-CGRP (mouse, rat) | 83651-90-5 | 99.8% | 3806.25 | 1 MG
SDP
Catalog No. 5000426280
Encompass_Preferred
α-CGRP (mouse, rat) is a neuropeptide (calcitonin gene-related peptide (CGRP)) mainly expressed in neuromuscular junction, which acts as a potent vasodilator. It can lead to a fall in blood pressure and an increase in heart rate by peripheral administration, and also relaxes colonic smooth muscle. This product has potential for use in cardiovascular, pro-inflammatory, migraine, and metabolic studies.
  • Potent vasodilator
  • Regulates innate lymphoid cell response
  • Regulates insulin secretion
  • Decreases mean arterial blood pressure
  • Increases heart rate and systemic vascular conduction
  • Plays a role in regulating Kainic acid-induced pyramidal cell death
  • Reduces frequency and severity of epileptic seizures in mice
  • Mitigates pathological damage in hippocampal regions
Catalog No. 50-004-26280 Supplier Medchemexpress LLC Supplier No. HYP02031MG
α-CGRP (mouse, rat) is a neuropeptide (calcitonin gene-related peptide (CGRP)) mainly expressed in neuromuscular junction, which acts as a potent vasodilator. It can lead to a fall in blood pressure and an increase in heart rate by peripheral administration, and also relaxes colonic smooth muscle. This product has potential for use in cardiovascular, pro-inflammatory, migraine, and metabolic studies.
  • Potent vasodilator
  • Regulates innate lymphoid cell response
  • Regulates insulin secretion
  • Decreases mean arterial blood pressure
  • Increases heart rate and systemic vascular conduction
  • Plays a role in regulating Kainic acid-induced pyramidal cell death
  • Reduces frequency and severity of epileptic seizures in mice
  • Mitigates pathological damage in hippocampal regions