Learn More
Eurofins Discoverx ChemiScreen CGRP1 Calcitonin Receptor-Like Receptor Stable Cell Line, 2 vials. (HTS172C)
Catalog No. 502006000
Encompass
The ChemiScreen CGRP1 Calcitonin Receptor-Like Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. calcitonin) based activation of CALCRL - RAMP1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CALCRL alongside RAMP1 modifying protein and endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of CALCRL, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-6000 Supplier Eurofins Discoverx Supplier No. HTS172C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen CGRP1 Calcitonin Receptor-Like Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. calcitonin) based activation of CALCRL - RAMP1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CALCRL alongside RAMP1 modifying protein and endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of CALCRL, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.